Learn More
StatLab CMV Placenta IHC Control Slides
SDP
Catalog No. 21033331
Change view
Click to view available options
Quantity:
15 Pk.
Catalog No. Quantity
21-033-331 15 Pk.
1 options
Catalog No. 21-033-331 Supplier StatLab Supplier No. CSCMV/15
Add to Cart
Add to Cart
Use CMV placenta control slides with confidence to evaluate stain quality and performance in histology laboratories
  • Slides consist of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • Includes 15 Unstained Slides per pack
Specifications
Clia Complexity Not Applicable
Type Control Slide
Quantity 15 Pk.
Form Solid