Learn More
Creative Biomart Recombinant CMV Pp150 Antigen
Supplier:  Creative Biomart CMV032
Encompass_Preferred
The E. coli derived recombinant protein contains the CMV Pp150 (UL32) immunodominant regions, 1011-1048 amino acids.
Catalog No. 50-208-4410