Learn More
Medchemexpress LLC Cmx-2043 (α-lipoic acid analogue) | 910627-26-8 | 98.8% | 406.52 | C16H26N2O6S2 | 10 MG
SDP
Catalog No. 5000229830
Encompass_Preferred
CMX-2043 is an α-lipoic acid analogue research compound with reported antioxidant and cytoprotective activity. It is used in preclinical studies to probe insulin receptor kinase and Akt-mediated signaling and is supplied in small, accurately weighed research quantities for laboratory use.
  • α-lipoic acid analogue with reported antioxidant activity.
  • Activates insulin receptor kinase and Akt-mediated signaling pathways.
  • Supplied in small, accurately weighed research quantities suitable for in vitro and preclinical studies.
  • High purity appropriate for research applications.
  • For laboratory research use only; not for human or clinical use.
Catalog No. 50-002-29830 Supplier Medchemexpress LLC Supplier No. HY11915210MG
CMX-2043 is an α-lipoic acid analogue research compound with reported antioxidant and cytoprotective activity. It is used in preclinical studies to probe insulin receptor kinase and Akt-mediated signaling and is supplied in small, accurately weighed research quantities for laboratory use.
  • α-lipoic acid analogue with reported antioxidant activity.
  • Activates insulin receptor kinase and Akt-mediated signaling pathways.
  • Supplied in small, accurately weighed research quantities suitable for in vitro and preclinical studies.
  • High purity appropriate for research applications.
  • For laboratory research use only; not for human or clinical use.