Learn More
LabStrong Corp CMX25 CLEANING SYRINGE 3P
Supplier:  LabStrong Corp CMX25LS
Encompass_Preferred
The LabStrong CMX25LS Cleaning Syringe is comparable to the Thermo Scientific™ Barnstead™ CMX25 Cleaning Syringe. The cleaning syringe is designed for use with EASYpure™ II LF, EASYpure™ UV, EASYpure™ UV/UF, NANOpure™ Infinity™ and NANOpure™ DIamond™ water purification systems. The syringe contains a cleaning solution specifically formulated to sanitize and flush away bacterial contaminants in Thermo Scientific™ Barnstead™ water purification systems. The LabStrong cleaning syringe provides outstanding quality at an economical price. The syringe features a clear barrel with bold scale markings, tapered plunger rod for ease of aspiration, and positive plunger rod stop. Thermo Scientific(tm) Barnstead(tm) Easypure(tm), Nanopure(tm) and Diamond(tm) are trademarks of Thermo Fisher Scientific Inc.
Catalog No. NC0910406