Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4609MSM4P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: 9E10.3. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: HL-60 cells or breast carcinoma. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: A synthetic peptide, corresponding to aa 408-439 (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc, coupled to KLH. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 62-64kDa.Chromosome: 8q24.21. Gene Symbol: MYC. Gene ID: 4609. Ensemble: ENSG00000136997. Uniprot: NP_002458.
Catalog No. 50-188-4979