Learn More
SPEX CERTIPREP LLC 3P Cyanide (complex)
Supplier:  SPEX CERTIPREP LLC 3P IS024
Encompass_Preferred
(NSI by Zeptometrix) 1000mg/L in Dilute NaOH. Derived from high purity Potassium ferricyanide, NIST traceable.
Catalog No. NC1601974
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.