Learn More
Medchemexpress LLC CN128 hydrochloride | 1335282-05-7 | 98.2% | 250 MG
SDP
Catalog No. 5000342080
Encompass_Preferred
CN128 hydrochloride (CN328) is an orally active and selective iron chelator primarily used in the research of β-thalassemia. It demonstrates high selectivity for iron(III) over other metals and has been shown to lack genetic toxicity.
  • Orally active and selective iron chelator
  • Used for research of β-thalassemia
  • Exhibits high selectivity for iron(III) over other metals
  • Lacks genetic toxicity
  • Demonstrates good oral bioavailability (82.6%) in rats
  • Well-tolerated in rats with no mortality at higher dose levels
  • Shows good iron scavenging efficacy in rats
Catalog No. 50-003-42080 Supplier Medchemexpress LLC Supplier No. 250MGHY131060
CN128 hydrochloride (CN328) is an orally active and selective iron chelator primarily used in the research of β-thalassemia. It demonstrates high selectivity for iron(III) over other metals and has been shown to lack genetic toxicity.
  • Orally active and selective iron chelator
  • Used for research of β-thalassemia
  • Exhibits high selectivity for iron(III) over other metals
  • Lacks genetic toxicity
  • Demonstrates good oral bioavailability (82.6%) in rats
  • Well-tolerated in rats with no mortality at higher dose levels
  • Shows good iron scavenging efficacy in rats