Learn More
Enzo Life Sciences Calcineurin inhibitory peptide (0.5mg). CAS: 148067-21-4
SDP
Supplier:  Enzo Life Sciences BMLPR1040500
Encompass_Preferred
Sequence is calcineurin (CaN) autoinhibitory domain (rat brain CaN aa 457-482). It inhibits calcineurin with an IC50 of 10 µM using [32P]myosin light chain as substrate. Inhibits Mn2+-stimulated calcineurin activity but has no effect on Ni2+-stimulated activity. Ki=4.4 µM with radiolabeled [Ala97]-RII (81-99) peptide as the phosphatase substrate. Alternative name: CN412. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1502