Learn More
Cayman Chemical (±)-Cannabichromene, Purity: ≥98%, CAS: 20675-51-8, 50 mg
SDP
Catalog No. 502935069
Encompass_Preferred
An analytical reference standard categorized as a phytocannabinoid; induces catalepsy, hypothermia, hypolocomotion, and antinociception in the mouse tetrad assay for cannabimimetic activity; intended for research and forensic applications
Catalog No. 50-293-5069 Supplier Cayman Chemical Supplier No. 2625250 MG
An analytical reference standard categorized as a phytocannabinoid; induces catalepsy, hypothermia, hypolocomotion, and antinociception in the mouse tetrad assay for cannabimimetic activity; intended for research and forensic applications