Learn More
Cayman Chemical Cannabicitran, Purity: ≥98%, CAS: 31508-71-1, 10 mg
SDP
Catalog No. 502916119
Encompass_Preferred
An analytical reference standard that is structurally similar to known phytocannabinoids; has been found in Cannabis and marijuana smoke condensate; intended for research and forensic applications
Catalog No. 50-291-6119 Supplier Cayman Chemical Supplier No. 3747510 MG
An analytical reference standard that is structurally similar to known phytocannabinoids; has been found in Cannabis and marijuana smoke condensate; intended for research and forensic applications