Learn More
Cayman Chemical Cannabidivarin, Purity: ≥98%, CAS: 24274-48-4, 250 mg
SDP
Catalog No. 502915492
Encompass_Preferred
An analytical reference standard categorized as a phytocannabinoid; considered an impurity in the commercial preparations of cannabidiol; intended for research and forensic applications
Catalog No. 50-291-5492 Supplier Cayman Chemical Supplier No. 36974250 MG
An analytical reference standard categorized as a phytocannabinoid; considered an impurity in the commercial preparations of cannabidiol; intended for research and forensic applications