Learn More
Cayman Chemical CnabIpIperIdIethnon 10mg
SDP
Catalog No. 502952522
Encompass_Preferred
A synthetic CB that binds to the CB receptors CB1 and CB2 with IC50 values of 591 and 968 nM, respectively; intended for forensic purposes and research applications
Catalog No. 50-295-2522 Supplier Cayman Chemical Supplier No. 1165510 MG
A synthetic CB that binds to the CB receptors CB1 and CB2 with IC50 values of 591 and 968 nM, respectively; intended for forensic purposes and research applications