Learn More
Cayman Chemical Cannabipiperidiethanone, Purity: ≥98%, CAS: 1345970-43-5, 5 mg
SDP
Catalog No. 502952189
Encompass_Preferred
A synthetic CB that binds to the CB receptors CB1 and CB2 with IC50 values of 591 and 968 nM, respectively; intended for forensic purposes and research applications
Catalog No. 50-295-2189 Supplier Cayman Chemical Supplier No. 116555 MG
A synthetic CB that binds to the CB receptors CB1 and CB2 with IC50 values of 591 and 968 nM, respectively; intended for forensic purposes and research applications