Learn More
Cayman Chemical Cannabis Flavonoids Mixture, 1 ea
SDP
Catalog No. 502915552
Encompass_Preferred
A mixture containing flavonoids in an N,N-dimethylacetamide solution; designed for use in method development and as a reference standard for these substances during routing analytical checks using HPLC and LC-MS instrumentation; intended for research and forensic applications
Catalog No. 50-291-5552 Supplier Cayman Chemical Supplier No. 378321 EA
A mixture containing flavonoids in an N,N-dimethylacetamide solution; designed for use in method development and as a reference standard for these substances during routing analytical checks using HPLC and LC-MS instrumentation; intended for research and forensic applications