Learn More
Cayman Chemical Canagliflozin, Purity: ≥98%, CAS: 842133-18-0, 10 mg
SDP
Catalog No. 502953701
Encompass_Preferred
An inhibitor of SGLT2 (IC50 = 2.2 nM) that less potently blocks SGLT1 (IC50 = 910 nM); lowers plasma glucose by lowering the renal threshold for glucose and increasing urinary glucose excretion in animals
Catalog No. 50-295-3701 Supplier Cayman Chemical Supplier No. 1157510 MG
An inhibitor of SGLT2 (IC50 = 2.2 nM) that less potently blocks SGLT1 (IC50 = 910 nM); lowers plasma glucose by lowering the renal threshold for glucose and increasing urinary glucose excretion in animals