Learn More
Cayman Chemical CnaglIflozIn 50mg
SDP
Catalog No. 502953669
Encompass_Preferred
An inhibitor of SGLT2 (IC50 = 2.2 nM) that less potently blocks SGLT1 (IC50 = 910 nM); lowers plasma glucose by lowering the renal threshold for glucose and increasing urinary glucose excretion in animals
Catalog No. 50-295-3669 Supplier Cayman Chemical Supplier No. 1157550 MG
An inhibitor of SGLT2 (IC50 = 2.2 nM) that less potently blocks SGLT1 (IC50 = 910 nM); lowers plasma glucose by lowering the renal threshold for glucose and increasing urinary glucose excretion in animals