Learn More
Cayman Chemical CnaglIflozIn-d4 1mg
SDP
Catalog No. 502932660
Encompass_Preferred
An internal standard for the quantification of canagliflozin by GC- or LC-MS
Catalog No. 50-293-2660 Supplier Cayman Chemical Supplier No. 252231 MG
An internal standard for the quantification of canagliflozin by GC- or LC-MS