Learn More
Aviva Systems Biology Corp CNBP Antibody - N-terminal region (ARP34815_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34815_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CNBP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CNBP. Alternate names: CNBP1, DM2, PROMM, RNF163, ZCCHC22, ZNF9ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55573