Learn More
Bioworld CNBR-ACTIVATED SEPAROPORE
Supplier:  Bioworld 201810033
Encompass
CNBR-ACTIVATED SEPAROPORE (AGAROSE) 4B: CNBR-Activated Separopore 4B Pre-activated medium for immobilization of ligands containing primary amines.; Reproducible, simple and reliable method for covalent immobilization of ligands containin...
Catalog No. 50-197-661