Learn More
Sigma Aldrich Fine Chemicals Biosciences CNBr Activated SepharoseR
Catalog No. 502904793
Encompass
CNBr-activated Sepharose(R) 4 fast flow is a well established pre-activated chromatography medium for coupling of large amino-containing ligands.CNBr-activated Sepharose(R) 4 fast flow is based on the well established Sepharose(R) fast flow platform. The resin is composed of cross-linked 4% agarose beads that has been pre-activated with cyanogen bromide. CNBr-activated Speharose 4 fast flow is designed for multipoint attachment of protein ligands containing amino groups.The preparation and use of affinity chromatography media by coupling biospecific ligands to CNBr-activated Sepharose(R) 4 fast flow is a widely used successful and well-documented approach with an easy rapid and efficient coupling procedure.CNBr-activated Sepharose(R) 4 fast flow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range CNBr-activated Sepharose(R) 4 fast flow meets industrial demands with
Catalog No. 50-290-4793 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17098101
CNBr-activated Sepharose(R) 4 fast flow is a well established pre-activated chromatography medium for coupling of large amino-containing ligands.CNBr-activated Sepharose(R) 4 fast flow is based on the well established Sepharose(R) fast flow platform. The resin is composed of cross-linked 4% agarose beads that has been pre-activated with cyanogen bromide. CNBr-activated Speharose 4 fast flow is designed for multipoint attachment of protein ligands containing amino groups.The preparation and use of affinity chromatography media by coupling biospecific ligands to CNBr-activated Sepharose(R) 4 fast flow is a widely used successful and well-documented approach with an easy rapid and efficient coupling procedure.CNBr-activated Sepharose(R) 4 fast flow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range CNBr-activated Sepharose(R) 4 fast flow meets industrial demands with