Learn More
Cayman Chemical CNDAC (hydrochloride), Purity: ≥98%, CAS: 134665-72-8, 50 mg
SDP
Catalog No. 502942515
Encompass_Preferred
A nucleoside derivative and an active metabolite of sapacitabine; induces chromosomal aberrations in UV41 cells lacking XPF at 1 µM; cytotoxic to BRCA1-/- UWB1.289 and BRCA2-/- PEO1 cells (IC50s = 9.9 and 10.2 nM, respectively)
Catalog No. 50-294-2515 Supplier Cayman Chemical Supplier No. 4067250 MG
A nucleoside derivative and an active metabolite of sapacitabine; induces chromosomal aberrations in UV41 cells lacking XPF at 1 µM; cytotoxic to BRCA1-/- UWB1.289 and BRCA2-/- PEO1 cells (IC50s = 9.9 and 10.2 nM, respectively)