Learn More
Cayman Chemical Cndesartn 100mg
SDP
Catalog No. 502894246
Encompass_Preferred
An AT1 receptor antagonist (Kis = 0.17, 0.12, and 0.12 nM for human AT1, rat AT1A, and rat AT1B recombinant receptors, respectively); selective for AT1 over AT2 receptors (Ki = 26,500 nM for the recombinant human AT2 receptor); inhibits angiotensin II-induced contraction of isolated rabbit aortic strips and increases in blood pressure in rats (ID50 = 0.033 mg/kg, i.v.)
Catalog No. 50-289-4246 Supplier Cayman Chemical Supplier No. 9003239100 MG
An AT1 receptor antagonist (Kis = 0.17, 0.12, and 0.12 nM for human AT1, rat AT1A, and rat AT1B recombinant receptors, respectively); selective for AT1 over AT2 receptors (Ki = 26,500 nM for the recombinant human AT2 receptor); inhibits angiotensin II-induced contraction of isolated rabbit aortic strips and increases in blood pressure in rats (ID50 = 0.033 mg/kg, i.v.)