Learn More
Cayman Chemical Candesartan-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1346604-70-3, 1 mg
SDP
Catalog No. 502936179
Encompass_Preferred
An internal standard for the quantification of candesartan by GC- or LC-MS
Catalog No. 50-293-6179 Supplier Cayman Chemical Supplier No. 254191 MG
An internal standard for the quantification of candesartan by GC- or LC-MS