Learn More
Cayman Chemical Candida genus qPCR Assay, 1 ea
SDP
Catalog No. 502943245
Encompass_Preferred
The primers and probes in the Candida genus kit are provided in tube 1 as a 20X concentrated working solution. This is a single assay that will detects 29 species of Candida including Candida albicans, Candida auris, Candida dubliniensis, Candida glabrata, Candida guiliermondii, Candida krusei, Candida lusitaniae, Candida parapsilosis, Candida tropicalis Organism Type: Fungi Controls (recommended): CrAssphage, Human RPP30-DNA Enzyme Master Mixes (recommended): Empirical qPCR mastermix, Standard qPCR enzyme mixture (Item No. 40705) Panels: Fungi, GI, Nail, Ophthalmology, UTI, Vaginal microbiota, Wound Colors: Cy5 (Candida genus), HEX (RPP30) Instruments: Any instrument
Catalog No. 50-294-3245 Supplier Cayman Chemical Supplier No. 406751 EA
The primers and probes in the Candida genus kit are provided in tube 1 as a 20X concentrated working solution. This is a single assay that will detects 29 species of Candida including Candida albicans, Candida auris, Candida dubliniensis, Candida glabrata, Candida guiliermondii, Candida krusei, Candida lusitaniae, Candida parapsilosis, Candida tropicalis Organism Type: Fungi Controls (recommended): CrAssphage, Human RPP30-DNA Enzyme Master Mixes (recommended): Empirical qPCR mastermix, Standard qPCR enzyme mixture (Item No. 40705) Panels: Fungi, GI, Nail, Ophthalmology, UTI, Vaginal microbiota, Wound Colors: Cy5 (Candida genus), HEX (RPP30) Instruments: Any instrument