Learn More
Cayman Chemical Candida M2 (Bio-Rad) Multiplexed qPCR Assay, 1 ea
SDP
Catalog No. 502943273
Encompass_Preferred
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X qPCR enzyme mastermix Organism Type: Fungi; Multiplexed Reaction Panels: Fungi, GI, Nail, Ophthalmology, UTI, Vaginal microbiota, Wound Colors: Cy5 (Candida auris), Cy5.5 (Candida lusitaniae), FAM (Candida dubliniensis), HEX (RPP30), TexasRED615 (Candida albicans) Instruments: Bio-Rad CFX96 or CFX384
Catalog No. 50-294-3273 Supplier Cayman Chemical Supplier No. 406771 EA
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X qPCR enzyme mastermix Organism Type: Fungi; Multiplexed Reaction Panels: Fungi, GI, Nail, Ophthalmology, UTI, Vaginal microbiota, Wound Colors: Cy5 (Candida auris), Cy5.5 (Candida lusitaniae), FAM (Candida dubliniensis), HEX (RPP30), TexasRED615 (Candida albicans) Instruments: Bio-Rad CFX96 or CFX384