Learn More
Aviva Systems Biology Corp CNDP1 Antibody - C-terminal region (ARP60609_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60609_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CNDP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CNDP1. Alternate names: CN1, CPGL2, HsT2308, MGC102737, MGC10825, MGC142072ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-102