Learn More
Aviva Systems Biology Corp CNDP2 Antibody - middle region (ARP57167_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57167_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CNDP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CNDP2. Alternate names: CN2, CPGL, FLJ10830, HsT2298, PEPAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53986