Learn More
Medchemexpress LLC CNDP2 Human sf9 H 10ug | 10ug
SDP
Catalog No. 5000185895
Encompass_Preferred
The CNDP2 protein is a catalyst that coordinates the oxidative cleavage of heme at the -methylene bridge carbon releasing carbon monoxide (CO) and generating biliverdin IXalpha This enzymatic process releases ferrous iron from the central heme iron chelate CNDP2 Protein Human (sf9 His) is the recombinant human-derived CNDP2 protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-85895 Supplier Medchemexpress LLC Supplier No. HYP7683710UG
The CNDP2 protein is a catalyst that coordinates the oxidative cleavage of heme at the -methylene bridge carbon releasing carbon monoxide (CO) and generating biliverdin IXalpha This enzymatic process releases ferrous iron from the central heme iron chelate CNDP2 Protein Human (sf9 His) is the recombinant human-derived CNDP2 protein expressed by Sf9 insect cells with C-His labeled tag