Learn More
AdipoGen Cnidilin
SDP
Supplier:  AdipoGen CDXC0727M005
Encompass_Preferred
Chemical. CAS 14348-22-2. Formula C17H16O5. MW 300.3. Cnidilin is a natural compound belonging to the coumarin family and normally isolated from Radix Angelicae dahuricae RAD, a traditional Chinese medicine TCM. RAD possesses anti-oxidant, anti-cancer and anti-inflammatory effects. Cnidilin has been described to show strong antiplatelet aggregation activity in vitro to be a GABAA receptor ligand and NO production inhibitor.
Catalog No. 50-256-1346