Learn More
Aviva Systems Biology Corp CNKSR2 Antibody - C-terminal region (ARP65739_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65739_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CNKSR2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human CNKSR2. Alternate names: CNK2, KIAA0902, KSR2, MAGUINApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-556