Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 1264MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: CNN1/832 + CALP. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: Myoepithelial cells in breast ducts or uterus. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human CNN1 protein (CNN1/832); Crude human uterus extract (CALP). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 34kDa.Chromosome: 19p13.2. Gene Symbol: CNN1. Gene ID: 1264. Ensemble: ENSG00000130176. Uniprot: XP_005259798.
Catalog No. 50-188-4619