Learn More
Aviva Systems Biology Corp CNNM4 Antibody - middle region (ARP49481_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49481_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CNNM4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CNNM4. Alternate names: ACDP4, KIAA1592ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60092