Learn More
Apexbio Technology LLC Clozapine N-oxide(CNO) dihydrochloride(Synonyms: CNO dihydrochloride, Clozapine N-oxide hydrochloride,Clozapine N-oxide dihydrochloride), 10mg, CAS: 2250025-93-3.
SDP
Catalog No. 5000060637
Encompass_Preferred
Clozapine N-oxide (CNO) dihydrochloride (CAS 2250025-93-3) is a synthetic metabolite derived from clozapine commonly utilized as an agonist for designer receptors exclusively activated by designer drugs (DREADDs) In neuroscience research CNO selectively binds and activates engineered muscarinic receptors particularly hM3Dq and hM4Di thereby influencing neuronal signaling upon penetrating the blood-brain barrier Due to its specificity CNO dihydrochloride serves as a valuable tool for dissecting neuronal circuits and elucidating their functional roles in behavioral studies
Catalog No. 50-000-60637 Supplier Apexbio Technology LLC Supplier No. B846610
Clozapine N-oxide (CNO) dihydrochloride (CAS 2250025-93-3) is a synthetic metabolite derived from clozapine commonly utilized as an agonist for designer receptors exclusively activated by designer drugs (DREADDs) In neuroscience research CNO selectively binds and activates engineered muscarinic receptors particularly hM3Dq and hM4Di thereby influencing neuronal signaling upon penetrating the blood-brain barrier Due to its specificity CNO dihydrochloride serves as a valuable tool for dissecting neuronal circuits and elucidating their functional roles in behavioral studies