Learn More
Aviva Systems Biology Corp CNOT2 Antibody - middle region (ARP32048_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32048_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CNOT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CNOT2. Alternate names: NOT2, CDC36, NOT2H, HSPC131ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54435