Learn More
Aviva Systems Biology Corp CNOT3 Antibody - N-terminal region (ARP34318_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34318_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CNOT3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CNOT3. Alternate names: NOT3, LENG2, NOT3HApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55342