Learn More
Aviva Systems Biology Corp CNOT4 Antibody - N-terminal region (ARP34307_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34307_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CNOT4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CNOT4. Alternate names: CLONE243, NOT4, NOT4HApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55335