Learn More
Bon Opus Biosciences Recombinant Human CNOT6 Protein - Source: E. coli
SDP
Catalog No. NC2733726
Encompass_Preferred
CCR4-NOT transcription complex subunit 6 is produced by our E.coli expression system and the target gene encoding C193-E340 is expressed with a 6His tag at the C-terminus. Accession#Q9ULM6. Formulation: Supplied as a 0.2 um filtered solution of 500mM PBS, 300mM NaCl, 10%Glycerol, pH7.4
Catalog No. NC2733726 Supplier Bon Opus Biosciences Supplier No. BP251050UG
CCR4-NOT transcription complex subunit 6 is produced by our E.coli expression system and the target gene encoding C193-E340 is expressed with a 6His tag at the C-terminus. Accession#Q9ULM6. Formulation: Supplied as a 0.2 um filtered solution of 500mM PBS, 300mM NaCl, 10%Glycerol, pH7.4