Learn More
Aviva Systems Biology Corp CNOT7 Antibody - middle region (ARP32607_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32607_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CNOT7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CNOT7. Alternate names: CAF1, hCAF-1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53984