Learn More
Aviva Systems Biology Corp Cnot8 Antibody - N-terminal region (ARP38510_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38510_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cnot8. It was validated on Western Blot. . Alternate names: 1500015I04Rik, 1810022F04Rik, AA536816, AU015770, AU043059ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-739