Learn More
MOLECULAR DEPOT LLC CNP-22 Human - 500 ug
Catalog No. 5000013467
Encompass_Preferred
CNP-22 (Human) is a synthetic peptide consisting of the sequence Gly-Leu-Ser-Lys-Gly-Cys-Phe-Gly-Leu-Lys-Leu-Asp-Arg-Ile-Gly-Ser-Met-Ser-Gly-Leu-Gly-Cys (Disulfide bond between Cys6-Cys22) The molecular weight of this peptide is 2197 6 g/mol and the CAS number is 131438-79-4 This product is for in vitro research use only This product is not for human or animal use
Catalog No. 50-000-13467 Supplier MOLECULAR DEPOT LLC Supplier No. MDP0614
CNP-22 (Human) is a synthetic peptide consisting of the sequence Gly-Leu-Ser-Lys-Gly-Cys-Phe-Gly-Leu-Lys-Leu-Asp-Arg-Ile-Gly-Ser-Met-Ser-Gly-Leu-Gly-Cys (Disulfide bond between Cys6-Cys22) The molecular weight of this peptide is 2197 6 g/mol and the CAS number is 131438-79-4 This product is for in vitro research use only This product is not for human or animal use