Learn More
Aviva Systems Biology Corp CNPY3 Antibody - middle region (ARP57943_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57943_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CNPY3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CNPY3. Alternate names: CAG4A, ERDA5, PRAT4A, TNRC5ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-286-418