Learn More
Medchemexpress LLC Cnqx | 115066-14-3 | C9H4N4O4 | 50 MG
SDP
Catalog No. 5000375841
Encompass_Preferred
CNQX is a potent and competitive AMPA/kainate receptor antagonist. It has IC50s of 0.3 μM for AMPA and 1.5 μM for kainate receptors, and also acts as a competitive non-NMDA receptor antagonist.
  • Potent and competitive AMPA/kainate receptor antagonist
  • IC50s of 0.3 μM for AMPA and 1.5 μM for kainate receptors
  • Acts as a competitive non-NMDA receptor antagonist
  • Blocks the expression of fear-potentiated startle in rats
  • Reversibly blocks Schaffer collateral and mossy fibre excitatory postsynaptic potential (EPSP)
  • Selectively reduces the amplitude of the monosynaptic component of DR-VRR
Catalog No. 50-003-75841 Supplier Medchemexpress LLC Supplier No. HY1506650MG
CNQX is a potent and competitive AMPA/kainate receptor antagonist. It has IC50s of 0.3 μM for AMPA and 1.5 μM for kainate receptors, and also acts as a competitive non-NMDA receptor antagonist.
  • Potent and competitive AMPA/kainate receptor antagonist
  • IC50s of 0.3 μM for AMPA and 1.5 μM for kainate receptors
  • Acts as a competitive non-NMDA receptor antagonist
  • Blocks the expression of fear-potentiated startle in rats
  • Reversibly blocks Schaffer collateral and mossy fibre excitatory postsynaptic potential (EPSP)
  • Selectively reduces the amplitude of the monosynaptic component of DR-VRR