Learn More
Medchemexpress LLC CNQX | 115066-14-3 | MFCD00069232 | C9H4N4O4 | 100 MG
SDP
Catalog No. 5000375276
Encompass_Preferred
CNQX (FG9065) is a potent and competitive AMPA/kainate receptor antagonist with IC50s of 0.3 μM and 1.5 μM, respectively. It functions as a competitive non-NMDA receptor antagonist and has been shown to block the expression of fear-potentiated startle in rats.
  • Potent and competitive AMPA/kainate receptor antagonist
  • IC50s of 0.3 μM for AMPA and 1.5 μM for kainate
  • Functions as a competitive non-NMDA receptor antagonist
  • Blocks expression of fear-potentiated startle in rats
  • Reversibly blocks Schaffer collateral and mossy fibre EPSP in hippocampal slices
  • Selectively reduces amplitude of monosynaptic component of DR-VRR in lumbar spinal segments
  • Decreases cocaine responses in a dose-dependent manner in rats
Catalog No. 50-003-75276 Supplier Medchemexpress LLC Supplier No. HY15066100MG
CNQX (FG9065) is a potent and competitive AMPA/kainate receptor antagonist with IC50s of 0.3 μM and 1.5 μM, respectively. It functions as a competitive non-NMDA receptor antagonist and has been shown to block the expression of fear-potentiated startle in rats.
  • Potent and competitive AMPA/kainate receptor antagonist
  • IC50s of 0.3 μM for AMPA and 1.5 μM for kainate
  • Functions as a competitive non-NMDA receptor antagonist
  • Blocks expression of fear-potentiated startle in rats
  • Reversibly blocks Schaffer collateral and mossy fibre EPSP in hippocampal slices
  • Selectively reduces amplitude of monosynaptic component of DR-VRR in lumbar spinal segments
  • Decreases cocaine responses in a dose-dependent manner in rats