Learn More
TARGETMOL CHEMICALS INC CNQX 25MG
SDP
Catalog No. 502576338
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CNQX (FG9065) is a competitive non-NMDA glutamate receptor antagonist (IC50s = 0.3 and 1.5 uM for AMPA and kainate receptors respectively)purity: 98%
Catalog No. 50-257-6338 Supplier TARGETMOL CHEMICALS INC Supplier No. T717825MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CNQX (FG9065) is a competitive non-NMDA glutamate receptor antagonist (IC50s = 0.3 and 1.5 uM for AMPA and kainate receptors respectively)purity: 98%