Learn More
Enzo Life Sciences CNQX (50mg). CAS: 115066-14-3
SDP
Supplier:  Enzo Life Sciences ALX550042M050
Encompass_Preferred
Potent, competitive kainate/quisqualate (non-NMDA) receptor antagonist. For the more soluble form see CNQX . disodium salt (ALX-550-283). Alternative name: 6-Cyano-7-nitroquinoxaline-2,3-dione, FG-9065. Purity: ≥98% (TLC). Appearance: Off-white to yellow powder. Solubility: Soluble in DMSO (10mg/ml) or dilute aqueous base. Long Term Storage: Ambient.
Catalog No. 50-200-9465