Learn More
Medchemexpress LLC HY-15066 5mg Medchemexpress, CNQX CAS:115066-14-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY150665MG
Encompass_Preferred
Medchemexpress, HY-15066 5mg CNQX CAS:115066-14-3 CNQX (FG9065) is a potent and competitive AMPA/kainate receptor antagonist with IC50s of 0.3 μM and 1.5 μM, respectively. CNQX is a competitive non-NMDA receptor antagonist. CNQX blocks the expression of fear-potentiated startle in rats[5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 502081677