Learn More
Medchemexpress LLC CNQX disodium | 479347-85-8 | C9H2N4Na2O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15066A100MG
Encompass_Preferred
CNQX disodium is a potent and competitive AMPA/kainate receptor antagonist. It acts as a competitive non-NMDA receptor antagonist and has demonstrated effectiveness in blocking the expression of fear-potentiated startle in rats.
  • Potent and competitive AMPA/kainate receptor antagonist.
  • Competitive non-NMDA receptor antagonist.
  • Blocks the expression of fear-potentiated startle in rats.
  • Reversibly blocks Schaffer collateral and mossy fibre excitatory postsynaptic potential (EPSP) in hippocampal slices.
  • Produces a selective and dose-dependent reduction in the amplitude of the monosynaptic component of the DR-VRR.
Catalog No. 50-003-75176