Learn More
Medchemexpress LLC CNQX disodium | 479347-85-8 | C9H2N4Na2O4 | 5 MG
SDP
Catalog No. 5000375297
Encompass_Preferred
CNQX disodium (FG9065 disodium) is a potent and competitive AMPA/kainate receptor antagonist. It blocks the expression of fear-potentiated startle in rats and is a competitive non-NMDA receptor antagonist. It is suitable for in vitro and in vivo research.
  • Potent and competitive AMPA/kainate receptor antagonist
  • Blocks expression of fear-potentiated startle
  • Competitive non-NMDA receptor antagonist
  • Suitable for neurological disease research
  • Inhibitor of ionotropic glutamate receptors
Catalog No. 50-003-75297 Supplier Medchemexpress LLC Supplier No. HY15066A5MG
CNQX disodium (FG9065 disodium) is a potent and competitive AMPA/kainate receptor antagonist. It blocks the expression of fear-potentiated startle in rats and is a competitive non-NMDA receptor antagonist. It is suitable for in vitro and in vivo research.
  • Potent and competitive AMPA/kainate receptor antagonist
  • Blocks expression of fear-potentiated startle
  • Competitive non-NMDA receptor antagonist
  • Suitable for neurological disease research
  • Inhibitor of ionotropic glutamate receptors