Learn More
Medchemexpress LLC CNQX disodium (FG9065 disodium) | 479347-85-8 | 100.0% | 276.12 g·mol⁻¹ | C9H2N4Na2O4 | 10 MG
SDP
Catalog No. 5000287263
Encompass_Preferred
CNQX disodium is the disodium salt of 6-cyano-7-nitroquinoxaline-2,3-dione, a potent and competitive AMPA/kainate (non-NMDA) ionotropic glutamate receptor antagonist used as a research reagent in electrophysiology and neuropharmacology. The disodium form is more water-soluble, facilitating preparation of aqueous solutions for in vitro and ex vivo assays.
  • Potent AMPA and kainate receptor antagonist with low-micromolar IC50s.
  • Water-soluble disodium salt for easy aqueous preparation.
  • High purity suitable for research applications.
  • Commonly used in electrophysiology and receptor pharmacology studies.
  • Supplied in small quantities appropriate for lab-scale experiments.
Catalog No. 50-002-87263 Supplier Medchemexpress LLC Supplier No. HY15066A10MG
CNQX disodium is the disodium salt of 6-cyano-7-nitroquinoxaline-2,3-dione, a potent and competitive AMPA/kainate (non-NMDA) ionotropic glutamate receptor antagonist used as a research reagent in electrophysiology and neuropharmacology. The disodium form is more water-soluble, facilitating preparation of aqueous solutions for in vitro and ex vivo assays.
  • Potent AMPA and kainate receptor antagonist with low-micromolar IC50s.
  • Water-soluble disodium salt for easy aqueous preparation.
  • High purity suitable for research applications.
  • Commonly used in electrophysiology and receptor pharmacology studies.
  • Supplied in small quantities appropriate for lab-scale experiments.