Learn More
Enzo Life Sciences CNQX . disodium salt (10mg). CAS: 115066-14-3 (non-salt)
SDP
Catalog No. 502009523
Encompass_Preferred
Water soluble, potent competitive kainate/quisqualate (non-NMDA) receptor antagonist. More soluble form of CNQX (ALX-550-042). Alternative name: 6-Cyano-7-nitroquinoxaline-2,3-dione . 2Na. Purity: ≥98% (TLC). Appearance: Rust red solid. Solubility: Soluble in water (5mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9523 Supplier Enzo Life Sciences Supplier No. ALX550283M010
Water soluble, potent competitive kainate/quisqualate (non-NMDA) receptor antagonist. More soluble form of CNQX (ALX-550-042). Alternative name: 6-Cyano-7-nitroquinoxaline-2,3-dione . 2Na. Purity: ≥98% (TLC). Appearance: Rust red solid. Solubility: Soluble in water (5mg/ml). Long Term Storage: -20°C.